APO455836480 Apollo Scientific Ltd

4-Bromo-3-chloro-2-fluoroaniline

  • MDL number MFCD04039280
  • Cas Number : 115843-99-7
  • Molecular Weight : 224.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-chloro-2-fluoroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-chloro-2-fluoroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart