APO455836489 Apollo Scientific Ltd

5-Fluoro-2-iodobenzyl alcohol

  • MDL number MFCD12031763
  • Cas Number : 877264-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Fluoro-2-iodophenyl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=57673

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=57673
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart