APO455836500 Apollo Scientific Ltd

2-Bromo-5-fluorobenzo[b]furan-3(2H)-one

  • MDL number MFCD28096612
  • Cas Number : 1823950-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-fluoro-1-benzofuran-3(2H)-one, 2-Bromo-2,3-dihydro-5-fluoro-3-oxo-1-benzofuran, 2-Bromo-5-fluorocoumaran-3-one
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64171

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64171
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart