APO455836503 Apollo Scientific Ltd

2'-Hydroxy-2,2,2,6'-tetrafluoroacetophenone

  • MDL number MFCD25541879
  • Cas Number : 1445995-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Fluoro-6-hydroxyphenyl)-2,2,2-trifluoroethan-1-one, 3-Fluoro-2-(trifluoroacetyl)phenol
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54609

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54609
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart