APO455836506 Apollo Scientific Ltd

2-(Difluoromethyl)benzyl alcohol

  • MDL number MFCD26728124
  • Cas Number : 1018678-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-(Difluoromethyl)phenyl]methanol, 2-(Hydroxymethyl)benzal fluoride
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59531

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=59531
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart