APO455836507 Apollo Scientific Ltd

4-Nitro-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD13185769
  • Cas Number : 50551-17-2
  • Molecular Weight : 219.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-5-nitrobenzotrifluoride, 4-Nitro-alpha,alpha,alpha-trifluoro-o-tolualdehyde
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-nitro-2-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-nitro-2-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart