APO455836508 Apollo Scientific Ltd

2-(Difluoromethyl)benzyl bromide

  • MDL number MFCD18791598
  • Cas Number : 1222556-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Bromomethyl)benzal fluoride, 1-(Bromomethyl)-2-(difluoromethyl)benzene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59534

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=59534
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart