APO455836513 Apollo Scientific Ltd

4-(Aminomethyl)-2-(trifluoromethyl)pyridine hydrochloride

  • MDL number MFCD12400889
  • Cas Number : 1159813-38-3
  • Molecular Weight : 212.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-aminomethyl-2-trifluoromethylpyridine-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-aminomethyl-2-trifluoromethylpyridine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart