APO455836523 Apollo Scientific Ltd

Methyl 4-methoxy-2-(trifluoromethyl)benzoate

  • MDL number MFCD06204092
  • Cas Number : 773875-67-5
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-48
Synonym 5-Methoxy-2-(methoxycarbonyl)benzotrifluoride, 4-(Methoxycarbonyl)-3-(trifluoromethyl)anisole
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-methoxy-2-trifluoromethylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-methoxy-2-trifluoromethylbenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart