APO455836525 Apollo Scientific Ltd

3-Bromo-6-fluoro-2-nitrotoluene

  • MDL number MFCD18632532
  • Cas Number : 1286734-82-4
  • Molecular Weight : 234.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-fluoro-3-methyl-2-nitrobenzene
Inchi Code 1S/C7H5BrFNO2/c1-4-6(9)3-2-5(8)7(4)10(11)12/h2-3H,1H3
Inchi Key PZZUMDDGTJMPQD-UHFFFAOYSA-N

Description

Synonym 6-Bromo-3-fluoro-2-methylnitrobenzene, 1-Bromo-4-fluoro-3-methyl-2-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-6-fluoro-2-nitrotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-6-fluoro-2-nitrotoluene
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart