APO455836527 Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethoxy)nitrobenzene

  • MDL number MFCD00042300
  • Cas Number : 124170-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Fluoro-2-nitro-4-(trifluoromethoxy)benzene, 3-Nitro-alpha,alpha,alpha,4-tetrafluoroanisole
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48678

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48678
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart