APO455836531 Apollo Scientific Ltd

4-(Difluoromethyl)benzyl bromide

  • MDL number MFCD18791603
  • Cas Number : 873373-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-4-(difluoromethyl)benzene, 4-(Bromomethyl)benzal fluoride
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59956

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=59956
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart