APO455836533 Apollo Scientific Ltd

2,4,6-Trifluorobenzyl methacrylate

  • MDL number MFCD28096613
  • Cas Number : 1820739-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,4,6-Trifluorobenzyl 2-methylprop-2-enoate
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64181

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64181
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart