APO455836536 Apollo Scientific Ltd

3-Fluoro-4-iodo-2-methylaniline

  • MDL number MFCD21364950
  • Cas Number : 329927-16-4
  • Molecular Weight : 251.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-4-iodo-2-methylaniline

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-4-iodo-2-methylaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart