APO455836540 Apollo Scientific Ltd

6-Bromo-3-chloro-2-fluorobenzyl bromide

  • MDL number MFCD28096619
  • Cas Number : 1820741-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-(bromomethyl)-4-chloro-3-fluorobenzene, 3-Chloro-alpha,6-dibromo-2-fluorotoluene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64175

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64175
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart