APO455836541 Apollo Scientific Ltd

2-Bromo-4-fluorobenzo[b]furan-3(2H)-one

  • MDL number MFCD28096620
  • Cas Number : 1823346-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-4-fluorocoumaran-3-one, 2-Bromo-4-fluoro-1-benzofuran-3(2H)-one
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64176

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=64176
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart