APO455836542 Apollo Scientific Ltd

6-Bromo-3-chloro-2-fluorobenzyl alcohol

  • MDL number MFCD24848703
  • Cas Number : 1449008-28-9
  • Molecular Weight : 239.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (6-Bromo-3-chloro-2-fluorophenyl)methanol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-3-chloro-2-fluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P280;P312
MSDS https://store.apolloscientific.co.uk/product/6-bromo-3-chloro-2-fluorobenzyl-alcohol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart