APO455836555 Apollo Scientific Ltd

6-Bromo-3-chloro-2-fluorobenzeneboronic acid

  • MDL number MFCD11111906
  • Cas Number : 1451392-87-2
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-3-chloro-2-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-3-chloro-2-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart