APO455836558 Apollo Scientific Ltd

6-Bromo-3-chloro-2-fluoroaniline, N-BOC protected

  • MDL number MFCD28096623
  • Cas Number : 1820705-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl (6-bromo-3-chloro-2-fluorophenyl)carbamate
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64236

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=64236
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart