APO455836560 Apollo Scientific Ltd

6-Bromo-3-chloro-2-fluorobenzylamine

  • MDL number MFCD28096624
  • Cas Number : 1499926-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (6-Bromo-3-chloro-3-fluorophenyl)methylamine
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=64238

Safety Information

signalword Danger
msds http://www.apolloscientific.co.uk/display_item.php?id=64238
pictograms GHS05;GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart