APO455836567 Apollo Scientific Ltd

Methyl 4-bromo-3-fluoro-2-hydroxybenzoate

  • MDL number MFCD28122826
  • Cas Number : 1807191-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-2-fluoro-6-(methoxycarbonyl)phenol, Methyl 4-bromo-3-fluorosalicylate
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=66642

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=66642
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart