APO455836569 Apollo Scientific Ltd

2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine

  • MDL number MFCD00153193
  • Cas Number : 33252-64-1
  • Molecular Weight : 208.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-190
Synonym 3-Nitro-5-(trifluoromethyl)pyridin-2-ol, 3-Nitro-5-(trifluoromethyl)pyridin-2(1H)-one, 6-Hydroxy-5-nitro-alpha,alpha,alpha-trifluoro-3-picoline
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-3-nitro-5-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-3-nitro-5-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart