APO455836570 Apollo Scientific Ltd

2-Hydroxy-5-nitro-3-(trifluoromethyl)pyridine

  • MDL number MFCD00276983
  • Cas Number : 99368-66-8
  • Molecular Weight : 208.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158
Synonym 5-Nitro-3-(trifluoromethyl)pyridin-2-ol, 5-Nitro-3-(trifluoromethyl)pyridin-2(1H)-one
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxy-5-nitro-3-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxy-5-nitro-3-trifluoromethylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart