APO455836584 Apollo Scientific Ltd

2-Fluoro-4-iodo-5-(methoxymethoxy)pyridine

  • MDL number MFCD22123918
  • Cas Number : 1034467-27-0
  • Molecular Weight : 283.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-iodo-5-methoxymethoxypyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-4-iodo-5-methoxymethoxypyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart