APO455836594 Apollo Scientific Ltd

Ethyl 2-[(2-fluorobenzoyl)amino]acetate

  • MDL number MFCD00793744
  • Cas Number : 304657-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-42
Synonym Ethyl (2-fluorobenzamido)acetate
Purity 97
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=3209

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=3209
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart