APO455836598 Apollo Scientific Ltd

2-Bromo-3-fluoro-6-nitroanisole

  • MDL number MFCD18071062
  • Cas Number : 1257535-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-4-fluoro-2-methoxynitrobenzene, 2-Bromo-1-fluoro-3-methoxy-4-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48303

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48303
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart