APO455836599 Apollo Scientific Ltd

3-Bromo-4-fluoro-2-methoxyaniline

  • MDL number MFCD18071064
  • Cas Number : 1257535-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Amino-2-bromo-3-fluoroanisole, 3-Bromo-4-fluoro-o-anisidine
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48305

Safety Information

signalword Warning
pcodes P261;P270;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48305
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart