APO455836600 Apollo Scientific Ltd

3-Bromo-2,4-difluoroaniline

  • MDL number MFCD12407090
  • Cas Number : 103977-79-3
  • Molecular Weight : 208.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-24-difluoroaniline

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-24-difluoroaniline
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart