APO455836603 Apollo Scientific Ltd

2-Bromo-3-chloro-5-fluoropyridine

  • MDL number MFCD13185350
  • Cas Number : 1214326-94-9
  • Molecular Weight : 210.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-chloro-5-fluoropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-chloro-5-fluoropyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart