APO455836604 Apollo Scientific Ltd

3-Chloro-5-fluoropyridine-2-carbonitrile

  • MDL number MFCD18255335
  • Cas Number : 950670-25-4
  • Molecular Weight : 156.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-fluoro-2-pyridinecarbonitrile
Inchi Code 1S/C6H2ClFN2/c7-5-1-4(8)3-10-6(5)2-9/h1,3H
Inchi Key BOPFMFYCILEZDH-UHFFFAOYSA-N

Description

Synonym 3-Chloro-5-fluoropicolinonitrile, 3-Chloro-2-cyano-5-fluoropyridine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-fluoropyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-fluoropyridine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart