APO455836605 Apollo Scientific Ltd

2-Fluoro-3-(trifluoromethyl)styrene

  • MDL number MFCD18909663
  • Cas Number : 1298094-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-3-vinylbenzotrifluoride, 1-Ethenyl-2-fluoro-3-(trifluoromethyl)benzene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48723

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48723
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart