APO455836608 Apollo Scientific Ltd

3-Chloro-5-fluoropyridine-2-carboxylic acid

  • MDL number MFCD13185822
  • Cas Number : 128073-01-8
  • Molecular Weight : 175.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144(dec.)
Synonym 3-Chloro-5-fluoropicolinic acid, 2-Carboxy-3-chloro-5-fluoropyridine
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-fluoropyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-fluoropyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart