APO455836617 Apollo Scientific Ltd

2,4-Difluoro-3-(trifluoromethyl)benzyl bromide

  • MDL number MFCD25541882
  • Cas Number : 1445995-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-2,4-difluoro-3-(trifluoromethyl)benzene, 3-(Bromomethyl)-2,6-difluorobenzotrifluoride
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54728

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54728
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart