APO455836622 Apollo Scientific Ltd

3-Amino-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD09955455
  • Cas Number : 49674-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Amino-5-cyanobenzotrifluoride, 5-Cyano-alpha,alpha,alpha-trifluoro-m-toluidine
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=59514

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=59514
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart