APO455836624 Apollo Scientific Ltd

Methyl 4-bromo-2-(trifluoromethoxy)benzoate

  • MDL number MFCD12761592
  • Cas Number : 933785-18-3
  • Molecular Weight : 299.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-bromo-2-(trifluoromethoxy)benzoate
Inchi Code 1S/C9H6BrF3O3/c1-15-8(14)6-3-2-5(10)4-7(6)16-9(11,12)13/h2-4H,1H3
Inchi Key QWWIUYXPVVJVNS-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-2-trifluoromethoxybenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-2-trifluoromethoxybenzoate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart