APO455836625 Apollo Scientific Ltd

4-Bromo-2-methoxybenzotrifluoride

  • MDL number MFCD13194486
  • Cas Number : 944805-63-4
  • Molecular Weight : 255.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-(trifluoromethyl)anisole, 5-Bromo-2-(trifluoromethyl)phenyl methyl ether, 4-Bromo-2-methoxy-1-(trifluoromethyl)benzene
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methoxybenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methoxybenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart