APO455836626 Apollo Scientific Ltd

3-Amino-4-(trifluoromethyl)benzonitrile

  • MDL number MFCD18072580
  • Cas Number : 1220630-83-0
  • Molecular Weight : 186.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-4-(trifluoromethyl)benzonitrile
Inchi Code 1S/C8H5F3N2/c9-8(10,11)6-2-1-5(4-12)3-7(6)13/h1-3H,13H2
Inchi Key CZYRYIJZODSVTK-UHFFFAOYSA-N

Description

Synonym 2-Amino-4-cyanobenzotrifluoride, 5-Cyano-alpha,alpha,alpha-trifluoro-o-toluidine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-trifluoromethylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-trifluoromethylbenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart