APO455836629 Apollo Scientific Ltd

4'-Bromo-2,2,2,2'-tetrafluoroacetophenone

  • MDL number MFCD19439730
  • Cas Number : 617706-18-0
  • Molecular Weight : 271.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(4-Bromo-2-fluorophenyl)-2,2,2-trifluoroethan-1-one
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2222-tetrafluoroacetophenone

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2222-tetrafluoroacetophenone
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart