APO455836630 Apollo Scientific Ltd

5-Fluoro-2-iodo-4-nitrotoluene

  • MDL number MFCD27937150
  • Cas Number : 1803811-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Fluoro-4-iodo-5-methyl-2-nitrobenzene, 2-Fluoro-5-iodo-4-methylnitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60655

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=60655
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart