APO455836632 Apollo Scientific Ltd

2-Fluoro-4-iodonitrobenzene 1g pack

  • MDL number MFCD18396989
  • Cas Number : 2996-31-8
  • Molecular Weight : 267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-4-iodo-1-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 118-118.5
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-4-iodonitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart