APO455836654 Apollo Scientific Ltd

3,3,3-Trifluoropropanal hydrate

  • MDL number MFCD18384868
  • Cas Number : 1309602-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,3,3-Trifluoropropionaldehyde hydrate (diol), 3,3,3-Trifluoropropionaldehyde hydrate, 3,3,3-Trifluoropropaldehyde hydrate
Density 1.3009
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 56-56.5/745mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48502

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48502
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart