APO455836655 Apollo Scientific Ltd

3-Bromo-2,4-difluoronitrobenzene

  • MDL number MFCD12545882
  • Cas Number : 103977-78-2
  • Molecular Weight : 237.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Synonym 2-Bromo-1,3-difluoro-4-nitrobenzene
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-24-difluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-24-difluoronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart