APO455836660 Apollo Scientific Ltd

2'-Amino-2,2,2-trifluoro-5'-(trifluoromethyl)acetophenone

  • MDL number MFCD18071074
  • Cas Number : 489429-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Trifluoroacetyl)-4-(trifluoromethyl)aniline, 1-[2-Amino-5-(trifluoromethyl)phenyl]-2,2,2-trifluoroethan-1-one, 4-Amino-3-(trifluoroacetyl)benzotrifluoride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48258

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48258
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart