APO455836661 Apollo Scientific Ltd

2-Bromo-5-fluoropyrimidine

  • MDL number MFCD09835164
  • Cas Number : 947533-45-1
  • Molecular Weight : 176.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-5-fluoro-1,3-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-fluoropyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-fluoropyrimidine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart