APO455836665 Apollo Scientific Ltd

4-Fluoro-3-nitrophenol

  • MDL number MFCD04038714
  • Cas Number : 2105-96-6
  • Molecular Weight : 157.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-5-hydroxynitrobenzene
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-nitrophenol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-nitrophenol
pictograms GHS05;GHS07
hstatements H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart