APO455836667 Apollo Scientific Ltd

4-(Trifluoromethyl)pyridine-3-boronic acid, pinacol ester

  • MDL number MFCD12198140
  • Cas Number : 1310405-06-1
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridine, 4,4,5,5-Tetramethyl-2-[4-(trifluoromethyl)pyridin-3-yl]-1,3,2-dioxaborolane
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoromethylpyridine-3-boronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoromethylpyridine-3-boronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart