APO455836673 Apollo Scientific Ltd

3-(Difluoromethyl)benzyl bromide

  • MDL number MFCD18791600
  • Cas Number : 1263178-51-3
  • Molecular Weight : 221.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-3-(difluoromethyl)benzene, alpha-Bromo-alpha',alpha'-difluoro-m-xylene, 3-(Bromomethyl)benzal fluoride
Purity 97
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-difluoromethylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-difluoromethylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart