APO455836677 Apollo Scientific Ltd

3-(Chloromethyl)-5-fluoropyridine hydrochloride

  • MDL number MFCD19687170
  • Cas Number : 1222556-83-3
  • Molecular Weight : 182.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloromethyl-5-fluoropyridine-hydrochloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloromethyl-5-fluoropyridine-hydrochloride
pictograms GHS05;GHS07
hstatements H302;H312;H314;H332
Availability details for
Quantity
  • Availability
  • Add to Cart