APO455836681 Apollo Scientific Ltd

3-Amino-5-fluoropyridine-2-carbonitrile

  • MDL number MFCD20259119
  • Cas Number : 1374115-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Amino-5-fluoropicolinonitrile, 3-Amino-2-cyano-5-fluoropyridine
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50309

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50309
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart