APO455836682 Apollo Scientific Ltd

5-Bromo-2-hydrazino-3-(trifluoromethyl)pyridine

  • MDL number MFCD22375529
  • Cas Number : 1374652-48-8
  • Molecular Weight : 256.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [5-Bromo-3-(trifluoromethyl)pyridin-2-yl]hydrazine, 5-Bromo-2-hydrazino-alpha,alpha,alpha-trifluoro-3-picoline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-hydrazino-3-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-hydrazino-3-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart